VWR®, ICP multi-element solution
Leverandør: VWR Chemicals
85036.180EA 3275 NOK
85036.180
VWR®, ICP multi-element solution
Standarder Kationstandarder Multi-Element Standards
ICP solution of eight elements in 7% nitric acid.
Cd 25 mg/L, Pb 5 mg/L, As 1,5 mg/L, Hg 15 mg/L, Mo 100 mg/L, Ni 500 mg/L, V 100 mg/L, Cu 1000 mg/L.
Sertifiseringer: USP 232 Oral dose
Traceable to SRM from NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.