VWR® ARISTAR®, ICP Multi-Element Solution diluted According USP 232 Oral dose, 8 Elements in 7% Nitric Acid
Leverandør: VWR Chemicals
85037.180EA 3275 NOK
85037.180
VWR® ARISTAR®, ICP Multi-Element Solution diluted According USP 232 Oral dose, 8 Elements in 7% Nitric Acid
Standarder Kationstandarder Multi-Element Standards
Eight elements in 7% nitric acid.
Cd 2,5 mg/L, Pb 5 mg/L, As 1,5 mg/L, Hg 1,5 mg/L, Mo 10 mg/L, Ni 50 mg/L, V 10 mg/L, Cu 100 mg/L in HNO₃ 7%.
Sertifiseringer: Traceable to SRM from NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Forpakning: USP232 oral dose