VWR® ARISTAR®, ICP Multi-Element Solution According USP232 Parental dose 6 Elements 100 mg/L in 15% Hydrochloric Acid
Leverandør: VWR Chemicals
85038.180EA 4475 NOK
85038.180
VWR® ARISTAR®, ICP Multi-Element Solution According USP232 Parental dose 6 Elements 100 mg/L in 15% Hydrochloric Acid
Standarder Kationstandarder Multi-Element Standards
Six elements: 100 mg/L each of Ir; Pt; Os; Rh; Pd; Ru in HCl 15%.
Sertifiseringer: Traceable to SRM from NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.