VWR® pH Buffer, Acetic Acid-Ammonium Acetate, pH 6,8, REAGENT
Leverandør: VWR Chemicals
85325.260EA 1850 NOK
85325.260
VWR® pH Buffer, Acetic Acid-Ammonium Acetate, pH 6,8, REAGENT
Buffere Buffere, pH
Ready to use test solution (TS).
  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available