Sykloheksan ≥99.7%, PESTINORM® Supra Trace for organic trace analysis

Leverandør: VWR Chemicals
Danger

Synonymer: Hexahydrobenzene, Hexamethylene

Filtered through a 0.2 µm filter, packaged under inert gas.

85385.320EA 1850 NOK
85385.320
Sykloheksan ≥99.7%, PESTINORM® Supra Trace for organic trace analysis
Sykloheksan
Formel: C₆H₁₂
MW: 84.16 g/mol
Kokepunkt: 81
Smeltepunkt: 7
Tetthet: 0.7785 g/cm³ (20 °C)
Flash punkt: –18 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003814
CAS-nummer: 110-82-7
EINECS: 203-806-2
FN: 1145
ADR: 3 II
REACH: 01-2119463273-41
Merck Indeks: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colourless liquid
Identity (IR) Conforms
Assay (calculated on anhydrous) Min. 99.7 %
Residue after evaporation Max. 0.0003 %(m)
Water Max. 0.01 M%
Colour Max. 10 APHA
GC/ECD dioxins, furans and PCB's Max. 5 ng/l
GC/ECD any pesticide (as Lindane) Max. 5 ng/l
GC/NPD any pesticide (as Parathion) Max. 10 ng/l
GC/ECD 1,2,4-TCB to deca-PCB(as Lindane) Max. 5 pg/ml
GC/ECD DCM to 1,2,4-TCB (as TCM) Max. 2.0 ng/ml
GC/MSD C10 to C40 (as Decane; 30-600amu) Max. 2.0 ng/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR