N,N-Dimetylacetamid 99.99-100.00%, PESTINORM® for headspace gas chromatography

Leverandør: VWR Chemicals
Danger

Synonymer: Acetic acid dimethylamide, Dimethyl Acetamide, DMAC

Filtered through a 0.2 µm filter, packaged under nitrogen.

85397.290EA 2310 NOK
85397.290
N,N-Dimetylacetamid 99.99-100.00%, PESTINORM® for headspace gas chromatography
N,N-Dimetylacetamid
Formel: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Kokepunkt: 166.1 °C (1013 hPa)
Smeltepunkt: –20 °C
Tetthet: 0.937 g/cm³ (20 °C)
Flash punkt: 70
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008686
CAS-nummer: 127-19-5
EINECS: 204-826-4
FN: 2810
ADR: 6.1 III
REACH: 01-2119459339-27
Merck Indeks: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity (GC) min. 99.8%
Identity (IR) Complying
Residue after evaporation max. 0.0003%w/w
Water (KF) max. 0.05%w/w
Color (APHA Pt-Co) max. 10
GC-HS (Each class 1 solvent) max. 1µg/g
GC-HS (Each class 2 solvent) max. 10µg/g
GC-HS (Each class 3 solvent) max. 50µg/g
GC-HS (DMF) max. 20µg/g
Transmittance (350 nm) Min. 98 %
Transmittance (400 nm) Min. 99 %
Headspace test for O.V.I's Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR