VWR® Rota®, pH indicator paper, 1 to 14
Leverandør: VWR Chemicals
85402.600EA 890 NOK
85402.600
VWR® Rota®, pH indicator paper, 1 to 14
Testpapir pH-indikatorstrips
Indicator paper for testing across the complete pH range.
  • Easy to use
  • Produced from high quality paper to give good colour matching of paper to printed calibration scale
Leveringsinformasjon: Supplied in plastic reels that ensure protection and long term stability. Reel of 5 m length, width: 7 mm.