VWR® Rota® Duo, pH indicator papers
Leverandør: VWR Chemicals
85403.600EA 890 NOK
85403.600 85404.600 85405.600 85406.600 85407.600 85408.600
VWR® Rota® Duo, pH indicator papers
Testpapir pH-indikatorstrips
Duo indicator papers show two different colours for each pH value.
  • Easy to use for qualitatively accurate readings
  • Produced from high quality paper to give good colour matching of paper to printed calibration scale
  • Narrow range papers with graduation intervals of 0.3 pH units on the colour scale
  • Wide range papers with graduation intervals up to 1 pH unit on the colour scale
Leveringsinformasjon: Supplied in plactic reels that ensure protection and long-term stability. Reel of 5 m length, width: 10 mm