VWR® Rota® Tri, pH indicator, 1 to 11
Leverandør: VWR Chemicals
85409.600EA 1175 NOK
85409.600
VWR® Rota® Tri, pH indicator, 1 to 11
Testpapir pH-indikatorstrips
These tri indicator papers show three different colours for each pH-value.
  • High precision due to the 3 colour scale
  • Easy to use
  • Produced from high quality paper to give good colour matching of paper to printed calibration scale
Tri paper has a hydrophobic barrier between the different indicators which ensures a distinctive separation of the colour bands.
Leveringsinformasjon: Supplied in plactic reels that ensure protection and long term stability. Reel of 6 m length, width: 14 mm