VWR® Dosatest, pH indicator strips for coloured samples
Leverandør: VWR Chemicals
85424.602EA 245 NOK
85424.602 85425.602 85426.602 85427.602 85428.602 85429.602 85430.602 85431.602 85432.602
VWR® Dosatest, pH indicator strips for coloured samples
Testpapir pH-indikatorstrips
Dosatest strips incorporate a colour scale and are ideal for determining pH values for coloured solutions.
  • Reliable even in coloured samples
  • Versatile and economic
  • Handle remains dry and clean whatever the sample
Due to the scale in the strip, any sample colour will have the same effect on the reference colour as on the reactive strip ensuring accurate readings even in coloured samples.
Invisible hydrophobic barrier above the top colour field on the strip stops capillary action of the test solution and prevents contact with the user.
Forpakning:More Product Information