Maursyre ( metansyre) 90% ≥90%, AnalaR NORMAPUR® ACS

Leverandør: VWR Chemicals
Danger

Synonymer: Aminic Acid, Methanoic acid

85495.290EA 635 NOK
85495.290
Maursyre ( metansyre) 90% ≥90%, AnalaR NORMAPUR® ACS
Maursyre ( metansyre)
Formel: HCO₂H
MW: 46.03 g/mol
Tetthet: 1.208 g/cm³ (20 °C)
Flash punkt: 48
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003297
CAS-nummer: 64-18-6
EINECS: 0
FN: 1779
ADR: 8 (3) II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay 89.0 - 91.0 %
Dilution test Passes test ACS
SO3 (Sulphite) Passes test
Colouration Max. 10 APHA
Acetic acid Max. 0.04 %
Evaporation residue Max. 20 ppm
Heavy metals (by ICP-MS) Max. 5 ppm
Cl (Chloride) Max. 10 ppm
NH4 (Ammonium) Max. 50 ppm
SO4 (Sulphate) Max. 20 ppm
Fe (Iron) Max. 5 ppm
Conforms to ACS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR