VWR® TITRINORM®, pH Buffers, for Karl Fischer Titrations
Leverandør: VWR Chemicals
85497.260EA 990 NOK
85497.260
VWR® TITRINORM®, pH Buffers, for Karl Fischer Titrations
Karl Fischer titranter og reagenser
The volumetric and coulometric reagents are buffered systems, stabilised around the ideal value between 5 to 7 ensuring a rapid and stoichiometric course of reaction. Buffering systems which neutralise and stabilise the pH during the reaction are suitable for this purpose.