Ammoniumsulfat ≥99.0% Reag. Ph. Eur.

Leverandør: VWR Chemicals

Synonymer: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

85509.290EA 800 NOK
85509.290
Ammoniumsulfat ≥99.0% Reag. Ph. Eur.
Ammonium sulphate
Formel: (NH₄)₂SO₄
Smeltepunkt: 336…339 °C
Tetthet: 1,78 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003391
CAS-nummer: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Indeks: 13,00559

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99.0 %
Appearance White crystals
pH (5 %) 4.5 - 6.0
Sulphated ash Max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR