Natriumhydroksid 6 mol/l (6 N) in aqueous solution for biopharmaceutical production

Leverandør: VWR Chemicals
Danger

85522.320EA 0 NOK
85522.320
Natriumhydroksid 6 mol/l (6 N) in aqueous solution for biopharmaceutical production
Natriumhydroksid
Formel: NaOH
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
FN: 1824
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Assay 5.9 --> 6.1 mol/l
Appearance Passes test
CO3 (as Na2CO3) Max. 1 %
Heavy metals (as Pb) Max. 5 ppm
Cl (Chloride) Max. 20 ppm
SO4 (Sulphate) Max. 30 ppm
Fe (Iron) Max. 3 ppm
Residual metal catalysts Unlikely to be present
Residual solvents (ICH Q3C) Excl.by manufact. process
Endotoxin Max. 2.0 EU/ml
Filtered through 0.2 μm Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR