VWR® ARISTAR®, Aluminium Standard Solutions for ICP-MS
Leverandør: VWR Chemicals
85548.180EA 1600 NOK
85548.180 457202A
VWR® ARISTAR®, Aluminium Standard Solutions for ICP-MS
Standarder Kationstandarder Aluminium Standards
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0,5% (typical)
  • Delivered with complete Certificate of Analysis
1000 ppm equivalent to 1000 mg/L
Sertifiseringer: Traceable to SRM from NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.