VWR® PESTINORM®, Cyclohexane, Secondary Reference Standard for GC
Leverandør: VWR Chemicals
85656.180EA 2925 NOK
85656.180
VWR® PESTINORM®, Cyclohexane, Secondary Reference Standard for GC
Standarder kromatografi standarder
A secondary GC solvent reference standard for various chromatographic and analytical applications.
  • Purity and the specific impurity profile is determined by GC-MS and reported in the CoA
  • Traceability data to specific current lot of primary standard
  • Completely synthetic and typically over 99,9% pure by GC-FID