VWR® PESTINORM®, 1,2-Dichloroethane, Secondary Reference Standard for GC
Leverandør: VWR Chemicals
85657.180EA 2925 NOK
85657.180
VWR® PESTINORM®, 1,2-Dichloroethane, Secondary Reference Standard for GC
Standarder kromatografi standarder
A secondary GC solvent reference standard for various chromatographic and analytical applications.
  • Purity and the specific impurity profile is determined by GC-MS and reported in the Certificate of Analysis
  • Traceability data to specific current lot of primary standard
  • Completely synthetic and typically over 99,9% pure by GC-FID