VWR® pH Buffer Solution According to Reag. Ph. Eur. 4000600
Leverandør: VWR Chemicals
85669.290EA 1100 NOK
85669.290
VWR® pH Buffer Solution According to Reag. Ph. Eur. 4000600
Buffere Buffere, pH
Contents: ammonium acetate R / water R / hydrochloric acid R1.
  • Ph Eur N 4000600
The reagent is specially intended for use in testing Ph Eur and monographs. This solution was produced according 4.1.1 of Ph Eur specifications using Ph Eur reagent quality components and water meeting the Ph Eur monograph requirement for purified water.
Bestillingsinformasjon: Normally manufactured to order. Please check with customer services.
Leveringsinformasjon: Delivery with certificate of analysis.