Sølvnitrat 4,791 g/l 0.0282 mol/l (0.0282 N) in aqueous solution

Leverandør: VWR Chemicals

Synonymer: Lunar caustic, Silver mononitrate, Nitric acid silver salt

85698.290EA 1550 NOK
85698.290
Sølvnitrat 4,791 g/l 0.0282 mol/l (0.0282 N) in aqueous solution
Sølvnitrat

1 ml = 1 mg Cl

Formel: AgNO₃
MW: 169.87 g/mol
Kokepunkt: 444
Smeltepunkt: 210
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003414
CAS-nummer: 7761-88-8
EINECS: 231-853-9
Merck Indeks: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR