Ascorbic acid test strips, Dosatest
Leverandør: VWR Chemicals
85701.601EA 515 NOK
85701.601
Ascorbic acid test strips, Dosatest
Miljø testkit
Semi-quantitative test strips for the determination of ascorbic acid.
  • Ideal for rapid and easy analysis directly on-site
  • No maintenance or calibration is required
Colour change: Yellow to green-blue
Leveringsinformasjon: Delivered as 100 test strips in a tube with instruction leaflet.