Cyanide test strips, Dosatest
Leverandør: VWR Chemicals
85703.601EA 920 NOK
85703.601
Cyanide test strips, Dosatest
Miljø testkit
Semi-quantitative test strips for the determination of cyanide.
  • Ideal for rapid and easy analysis of cyanide directly on-site
  • No maintenance or calibration is required
Colour change: White to violet
Leveringsinformasjon: Delivered as 100 test strips in a tube, additional regaents, instruction leaflet in a box