Phosphate test strips, Dosatest
Leverandør: VWR Chemicals
85704.601EA 695 NOK
85704.601
Phosphate test strips, Dosatest
Miljø testkit
Semi-quantitative test strips for the determination of phosphate.
  • Ideal for rapid and easy analysis of phosphate directly on-site
  • No maintenance or calibration is required
Colour change: White to blue-green
Leveringsinformasjon: Delivered as 100 test strips in a tube with instruction leaflet.