Molybdenum test strips, Dosatest
Leverandør: VWR Chemicals
85706.601EA 1050 NOK
85706.601
Molybdenum test strips, Dosatest
Miljø testkit
Semi-quantitative test strips for the determination of molybdenum.
  • Ideal for rapid and easy analysis of molybdenum directly on-site
  • No maintenance or calibration is required
Colour change: White to green.
Leveringsinformasjon: Delivered as 100 test strips in a tube, additional regaents, instruction leaflet in a box