Formaldehyde Test Strips, Dosatest
Leverandør: VWR Chemicals
85707.601EA 810 NOK
85707.601
Formaldehyde Test Strips, Dosatest
Miljø testkit
Semi-quantitative test strips for the determination of formaldehyde.
  • Ideal for rapid and easy analysis directly on-site
  • No maintenance or calibration is required
Colour change: Beige to blue-violet
Leveringsinformasjon: Delivered as 100 test strips in a tube, additional regaents, instruction leaflet in a box