Sulphate test strips, Dosatest, VWR®
Leverandør: VWR Chemicals
85708.601EA 490 NOK
85708.601
Sulphate test strips, Dosatest, VWR®
Miljø testkit
Semi-quantitative test strips for the determination of sulphate.
  • Ideal for rapid and easy analysis of sulphate directly on-site
  • No maintenance or calibration is required
Colour change: Red to yellow
Leveringsinformasjon: Delivered as 100 test strips in a tube with instruction leaflet.
NB: If the sample solution has a pH value of 4 to 8, the reaction will take place without interferences. Strongly acid solutions have to be buffered with sodium acetate, and alkaline with tartaric acid.