Glucose Test Strips, Dosatest
Leverandør: VWR Chemicals
85714.601EA 465 NOK
85714.601
Glucose Test Strips, Dosatest
Miljø testkit
Semi-quantitative test strips for the determination of glucose.
  • Ideal for rapid and easy analysis of glucose directly on-site
  • No maintenance or calibration is required
Colour change: Yellow to blue-green
Leveringsinformasjon: Delivered as 100 test strips in a tube with instruction leaflet.