VWR® ARISTAR®, Ion chromatography anion multi-element standard solution, Certified Reference Material
Leverandør: VWR Chemicals
85783.154EA 1975 NOK
85783.154
VWR® ARISTAR®, Ion chromatography anion multi-element standard solution, Certified Reference Material
Standarder Anionstandarder Multi-Element Anioinic Standards
This certified reference material is produced using a high purity starting material, acid from sub-boiling and 18 MOhm deionized water and filtered through a 0.2 micron filter.
10 mg/kg each of: F⁻, Cl⁻, Br-, NO₃⁻, PO₄³⁻, SO₄²⁻ in H2O
Sertifiseringer: CRM ISO 17034 and ISO 17025. Traceable to NIST.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Lagringstemp Ambient