Metanol ≥99.9%, HiPerSolv CHROMANORM® for UHPLC-MS

Leverandør: VWR Chemicals
Danger

Synonymer: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

Filtered through a 0.1 micron filter.

85802.290EA 1330 NOK
85802.290
Metanol ≥99.9%, HiPerSolv CHROMANORM® for UHPLC-MS
Metanol
Formel: CH₃OH
MW: 32.04 g/mol
Kokepunkt: 64.6 °C (1013 hPa)
Smeltepunkt: –98 °C
Tetthet: 0.7918 g/cm³ (20 °C)
Flash punkt: 11 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004595
CAS-nummer: 67-56-1
EINECS: 200-659-6
FN: 1230
ADR: 3 (6.1) II
REACH: 01-2119433307-44
Merck Indeks: 13,05984

Bestill nå

Spesifikasjon Testresultater

Assay (GC) min. 99.9%
Identity (IR) complying
Clear colorless liquid complying
Color (APHA Pt-Co) max. 10
Water(KF) max. 0.01% w/w
Residue after evaporation max. 0.7 ppm
Acidity max. 0.001 meq/g
Alkalinity max. 0.0002 meq/g
Aluminium (Al) max. 5 ppb
Calcium (Ca) max. 5 ppb
Iron (Fe) max. 5 ppb
Potassium (K) max. 5 ppb
Magnesium (Mg) max. 5 ppb
Sodium (Na) max. 25 ppb
Fluorescence 254/450 nm (as Quinine) max. 0.5 ppb
Fluorescence 365/450 nm (as Quinine) max. 0.5 ppb
UHPLC/MS ESI/APCI+ (as Reserpine) max. 2 ppb
UHPLC/MS ESI/APCI- (as Reserpine) max. 10 ppb
PEG Signal-to-noise ratio max. 50
UHPLC-UV Gradient (230 nm) max. 2.0 mAU
UHPLC-UV Gradient (254 nm) max. 0.5 mAU
UV Absorbance 210nm max. 0.50 AU
UV Absorbance 215 nm max. 0.40 AU
UV Absorbance 220 nm max. 0.20 AU
UV Absorbance 230 nm max. 0.10 AU
UV Absorbance 254 nm max. 0.01 AU
UV Absorbance 260 nm max. 0.005 AU
UV Absorbance 280 nm max. 0.005 AU
Filtered through 0.1 µm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR