Natriumklorid 0,5 mol/l 0,5 N aqueous solution, AVS TITRINORM Volumetric

Leverandør: VWR Chemicals

Synonymer: Saline, NaCl, Sea salt, Halite

85812.290EA 351 NOK
85812.290
Natriumklorid 0,5 mol/l 0,5 N aqueous solution, AVS TITRINORM Volumetric
Natriumklorid
Formel: NaCl
MW: 58.44 g/mol
Kokepunkt: 1413
Smeltepunkt: 801
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003477
CAS-nummer: 7647-14-5
Merck Indeks: 14,08599

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR