N,N-Dimetylacetamid, anhydrous (max. 0.005% H₂O)

Leverandør: VWR Chemicals
Danger

Synonymer: Acetic acid dimethylamide, Dimethyl Acetamide, DMAC

85817.230EA 1200 NOK
85817.230
N,N-Dimetylacetamid, anhydrous (max. 0.005% H₂O)
N,N-Dimetylacetamid
Formel: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Kokepunkt: 166.1 °C (1013 hPa)
Smeltepunkt: –20 °C
Tetthet: 0.937 g/cm³ (20 °C)
Flash punkt: 70 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008686
CAS-nummer: 127-19-5
EINECS: 204-826-4
FN: 2810
ADR: 6.1 III
REACH: 01-2119459339-27
Merck Indeks: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (GC, on anhydrous basis) 99.8-100 %
Acidity 0-0.0005 0-0.001 %
Residue after evaporation 0-0.001 % w/w
Water (KF) 0-0.005 % w/w


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR