Tris-(hydroksymetyl)aminometan hydroklorid ≥99.0% for biofarmasøytisk produksjon

Leverandør: VWR Chemicals

85827.297EA 3150 NOK
85827.297 85827.366 85827.468
Tris-(hydroksymetyl)aminometan hydroklorid ≥99.0% for biofarmasøytisk produksjon
Tris-(hydroksymetyl)aminometan hydroklorid
Formel: NH₂C(CH₂OH)₃·HCl
Smeltepunkt: 150…152 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012590
CAS-nummer: 1185-53-1
EINECS: 214-684-5
Merck Indeks: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99,0 %
Appearance White crystalline powder
Dnases Not detected
Identification by IR Conforms to structure
Proteases Not detected
Rnases Not detected
Solubility (1 mol/l) Clear and complete
pH (0,5 mol/l; 25°C) 3,5 -- 5,0
pKa (25°C) 8,0 -- 8,4
Ignition residue Max. 0,1 %
Insoluble substances Max. 50 ppm
Loss on drying (105°C) Max. 0,5 %
Ca (Calcium) Max. 5 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 5 ppm
Mg (Magnesium) Max. 5 ppm
Mn (Manganese) Max. 5 ppm
Pb (Lead) Max. 5 ppm
Zn (Zinc) Max. 5 ppm
Absorbance (260 nm) (1 mol/l) Max. 0,05
Absorbance (280 nm) (1 mol/l) Max. 0,05
Bioburden Max. 100 CFU/g
Endotoxin Max. 0.1 EU/mg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR