Etanol Eurodenatured ≥80%, TechniSolv® (1:1:1)

Leverandør: VWR Chemicals

85828.440EA 1850 NOK
85828.440
Etanol Eurodenatured ≥80%, TechniSolv® (1:1:1)
Etanol Eurodenatured
Formel: H₃CCH₂OH
MW: 46.07 g/mol
CAS-nummer: 64-17-5
FN: 1170
ADR: 3 II

Bestill nå

Spesifikasjon Testresultater

Assay (V/V) min. 78.0%
Denatonium benzoate (ppm) 9.3 - 10.8 ppm
Methyl ethyl ketone (V/V) 0.78 - 0.91 %
2-Propanol (V/V) 0.78 - 0.91%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR