Diklormetan ≥99.9% stabilised, PESTINORM® for MOSH/MOAH analysis

Leverandør: VWR Chemicals
Warning

Synonymer: Methylene chloride

Stabilisering: Stabilised with amylene
Filtered through a 0.2 µm filter, packaged under inert gas.

85958.320EA 1900 NOK
85958.320
Diklormetan ≥99.9% stabilised, PESTINORM® for MOSH/MOAH analysis
Diklormetan

Mineral oils are almost universally present in our environment. The main compounds of interest are mineral oil saturated hydrocarbons (MOSH) and mineral oil aromatic hydrocarbons (MOAH).
Both types are easily absorbed by the human body from food and can accumulate in body fat and in some organs.


  • Specifically designed and tested for the MOSH/MOAH analysis
  • Tested and guaranteed by HPLC/UV – GC-FID and GC-MS to have less than 3 ng/ml, and by GC-ECD less than 3 pg/ml of any peak in the full range from C10 to C50 of MOSH/MOAH impurities

Formel: CH₂Cl₂
MW: 84.93 g/mol
Kokepunkt: 39.8 °C (1013 hPa)
Smeltepunkt: –95 °C
Tetthet: 1.322 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000881
CAS-nummer: 75-09-2
EINECS: 200-838-9
FN: 1593
ADR: 6.1 III
REACH: 01-2119480404-41
Merck Indeks: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless liquid
Identity (IR) Complies
Color (APHA, Pt-Co) max. 10
Assay (GC, on anhydrous basis) min. 99.9 %
MOSH Test by GC/FID max. 5 ng/ml
MOAH Test by LC/UV max. 5 ng/ml
GC/ECD 1,2,4-TCB to Deca-PCB (as Lindane) max. 3 pg/ml
Water (KF) max. 0.02 % w/w
Residue after evaporation max. 0.0003 % w/w


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR