Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103

Leverandør: VWR Chemicals


Stabilisering: Stabilised with mercury (II) iodide 0,01 %

85964.180EA 1725 NOK
85964.180
Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
Starch

Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: (C₆H₁₀O₅)n
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00082026
CAS-nummer: 9005-25-8
EINECS: 232-679-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR