Svovelsyre 1 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1086804

Leverandør: VWR Chemicals
Warning

85973.290EA 1550 NOK
85973.290
Svovelsyre 1 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1086804
Svovelsyre

Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: H₂SO₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064589
CAS-nummer: 7664-93-9
EINECS: 231-639-5
FN: 3264
ADR: 8,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR