Tioacetamid 4% in aqueous solution Reag. Ph. Eur. 1089602

Leverandør: VWR Chemicals
Danger

85975.290EA 5000 NOK
85975.290 85975.180 85975.260
Tioacetamid 4% in aqueous solution Reag. Ph. Eur. 1089602
Tioacetamid

Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: CH₃CSNH₂
Kokepunkt: ~100 °C (1013 hPa)
Tetthet: 1 g/cm³ (20 °C)
MDL nummer: MFCD00008070
CAS-nummer: 62-55-5
EINECS: 200-541-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2000e62db991c85340c257ac6d64ca42c3551a&vid=18e76b2cdee9ee22a2618ec371858a498f218457&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR