Dodecyltrimethylammonium bromide ≥99%, GPR RECTAPUR®

Leverandør: VWR Chemicals

Synonymer: Dodecyl trimethyl ammonium bromide, (1-Dodecyl)trimethylammonium bromide

85982.130EA 1175 NOK
85982.130
Dodecyltrimethylammonium bromide ≥99%, GPR RECTAPUR®
Dodecyltrimethylammonium bromide
Formel: C₁₅H₃₄BrN
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011767
CAS-nummer: 1119-94-4
FN: 2811
ADR: 6.1,II

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99 %
IR Spectrum Conforms to structure
Colouration (10 %; water) Max. 100 APHA
Solubility in water (10 %) Clear to slightly hazy


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR