N,N-Dimetyl-p-toluidine ≥98.0% (by GC), GPR RECTAPUR®

Leverandør: VWR Chemicals
Danger

85986.230EA 805 NOK
85986.230 85986.290 85986.460
N,N-Dimetyl-p-toluidine ≥98.0% (by GC), GPR RECTAPUR®
N,N-Dimetyl-p-toluidine
Formel: C₉H₁₃N
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008316
CAS-nummer: 99-97-8
FN: 2810
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance Clear pale yellow to amber liquid
Assay (GC) min. 98,0 %
Water max. 0,5 %
Density (d20°C/4°C) 0,935 – 0,937
Identity (IR) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR