Methyl red hydrochloride in ACS Reagent grade water, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

Synonymer: 2-(4-Dimethylaminophenylazo)benzoic acid hydrochloride

M-Clarity™ quality level = MQ200

861235-25G
861235-25GEA 393 NOK
861235-25G
Methyl red hydrochloride in ACS Reagent grade water, Sigma-Aldrich®
Methyl red hydrochloride
Formel: C₁₅H₁₆ClN₃O₂
Smeltepunkt: 175 °C
MDL nummer: MFCD00012614
CAS-nummer: 63451-28-5
EINECS: 264-190-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a50d91aa170afec176b5644b14a7252710cb2486&vid=918600653c77053735d47841c9e53d3cba4389d9&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Colour) Light Blue to Very Dark Blue and Light Purple to Very Dark Purple
Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Clarity of Solution
c = 0.1%; Ethanol
Conforms
Visual Interval
pH 4.2
Pink
Visual Interval
pH 5.5
Orange
Visual Interval
pH 6.2
Yellow
Meets ACS Requirements Current ACS Specification

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR