Kaliumdihydrogenortofosfat 1/15 mol/l (1/15 M), AVS TITRINORM stock solution_buffer

Leverandør: VWR Chemicals

Synonymer: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

86185.290EA 605 NOK
86185.290
Kaliumdihydrogenortofosfat 1/15 mol/l (1/15 M), AVS TITRINORM stock solution_buffer
Kaliumdihydrogenortofosfat
Formel: KH₂PO₄
MW: 136.09 g/mol
Kokepunkt: 100
Smeltepunkt: 253
MDL nummer: MFCD00011401
CAS-nummer: 7778-77-0
Merck Indeks: 13,07744

Bestill nå

Spesifikasjon Testresultater

Potassium phosphate, monobasic 0.0664 – 0.0670 mol/l
The solution is produced by very exact volumetric or gravimetric procedures. A final analysis is not carried out.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR