Natriumhydroksid 0.02 mol/l (0.02 N), AVS TITRINORM

Leverandør: VWR Chemicals

Synonymer: Caustic soda

86188.260EA 275 NOK
86188.260
Natriumhydroksid 0.02 mol/l (0.02 N), AVS TITRINORM
Natriumhydroksid
Formel: NaOH
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR