Fenolftalein 1%, TECHNICAL

Leverandør: VWR Chemicals
Danger

Synonymer: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

8626.1000EA 420 NOK
8626.1000
Fenolftalein 1%, TECHNICAL
Fenolftalein

  • Transition range: pH 8,2-9,8

Formel: C₂₀H₁₄O₄
Kokepunkt: 82 °C (1013 hPa)
Tetthet: 0,79 g/cm³ (20 °C)
Flash punkt: 12 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005913
CAS-nummer: 77-09-8
EINECS: 201-004-7
FN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Sensitivity Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR