Sølv 1.000 mg/l i fortynnet salpetersyre AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
86659.260EA 364 NOK
86659.260 86659.180
Sølv 1.000 mg/l i fortynnet salpetersyre AVS TITRINORM standard til AAS
Standarder Kationstandarder Silver Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ISO 17025 accredited facilities
  • Supplied with Certificate of Analysis, real value ±0,3%