Gull 1.000 mg/l Au in 2% saltsyre AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
86662.260EA 5000 NOK
86662.260 86662.180
Gull 1.000 mg/l Au in 2% saltsyre AVS TITRINORM standard til AAS
Standarder Kationstandarder Gold Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%