Boron 1.000 mg/l B i vandig AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
86663.260EA 329 NOK
86663.260 86663.180
Boron 1.000 mg/l B i vandig AVS TITRINORM standard til AAS
Standarder Kationstandarder Boron Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%