Kobalt 1.000 mg/l Co in dil. salpetersyre AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
86670.260EA 357 NOK
86670.260 86670.180
Kobalt 1.000 mg/l Co in dil. salpetersyre AVS TITRINORM standard til AAS
Standarder Kationstandarder Cobalt Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%