Germanium 1.000 mg/l Ge in 5% salpetersyre with hydrofluoric acid (max. 1%) AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
86680.180EA 510 NOK
86680.180 86680.260
Germanium 1.000 mg/l Ge in 5% salpetersyre with hydrofluoric acid (max. 1%) AVS TITRINORM standard til AAS
Standarder Kationstandarder Germanium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%