Holmium 1.000 mg/l i fortynnet salpetersyre AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
86683.180EA 605 NOK
86683.180 86683.260
Holmium 1.000 mg/l i fortynnet salpetersyre AVS TITRINORM standard til AAS
Standarder Kationstandarder Holmium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%