Indium 1.000 mg/l i fortynnet salpetersyre AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
86684.180EA 890 NOK
86684.180 86684.260
Indium 1.000 mg/l i fortynnet salpetersyre AVS TITRINORM standard til AAS
Standarder Kationstandarder Indium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%