Iridium 1.000 mg/l i 10% saltsyre AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
86685.180EA 2800 NOK
86685.180 86685.260
Iridium 1.000 mg/l i 10% saltsyre AVS TITRINORM standard til AAS
Standarder Kationstandarder Iridium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%